Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-iodoanisole, 98%

$90.05 - $341.72

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-iodoanisole
CAS 52807-27-9
Molecular Formula C7H6ClIO
Formula Weight 268.48
MDL Number MFCD00052858
Synonym 5-Chloro-2-methoxyiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1384014

 
alfa aesar™
A1384014
25g Each for $341.72

AAA1384006

 
alfa aesar™
A1384006
5g Each for $90.05
Description & Specifications

Specifications

Formula Weight 268.48
Chemical Name or Material 4-Chloro-2-iodoanisole
Assay 98%
Molecular Formula C7H6ClIO
MDL Number MFCD00052858
CAS 52807-27-9
Synonym 5-Chloro-2-methoxyiodobenzene
Boiling Point 133° to 135°C (15mmHg)
Melting Point 44° to 48°C
Quantity 25g

Applications
An independent synthesis of 4-chloro-2-iodoanisole has shown that this is chlorinating agents, brominating agents and iodinating agents. 4-Chloro-2-iodoanisole (2) was demethylated and subsequently alkylated with 4-chloro-2-fluorobenzyl bromide, an aryl Grignard reagent was formed with it.

Solubility
Soluble in methanol.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.