Please login to your online account to display your discounted pricing

4-chloro-2-iodoaniline, +98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396840010

 View more versions of this product

Catalog No. AC396840010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥98.0% (GC)
Formula Weight 253.47
Quantity 1g
Color White to off-white
Melting Point 37° to 42°C
CAS Min % 98.0
CAS Max % 100.0
Molecular Formula C6H5ClIN
CAS 63069-48-7
Packaging Glass bottle
Chemical Name or Material 4-chloro-2-iodoaniline
Assay Percent Range 98+%
MDL Number MFCD01863737
Infrared Spectrum Authentic
Linear Formula ClC6H3(I)NH2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

Specifications

Chemical Name or Material 4-chloro-2-iodoaniline
CAS 63069-48-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0% (GC)
Assay Percent Range 98+%
Molecular Formula C6H5ClIN
Linear Formula ClC6H3(I)NH2
Formula Weight 253.47
MDL Number MFCD01863737
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

SDS & Safety Documents