Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-hydroxypyridine, 98%

$115.14 - $375.58

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-hydroxypyridine
CAS 40673-25-4
Molecular Formula C5H4ClNO
Formula Weight 129.55
MDL Number MFCD03093022
Synonym 4-Chloro-2(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2925003

  SDS
alfa aesar™
H2925003
1g Each for $375.58

AAH29250MD

  SDS
alfa aesar™
H29250MD
250mg Each for $115.14
Description & Specifications

Specifications

Formula Weight 129.55
Chemical Name or Material 4-Chloro-2-hydroxypyridine
Assay 98%
Molecular Formula C5H4ClNO
MDL Number MFCD03093022
CAS 40673-25-4
Synonym 4-Chloro-2(1H)-pyridone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 177° to 179°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.