Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorotoluene, 99%

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-fluorotoluene
CAS 452-75-5
Molecular Formula C7H6ClF
Formula Weight 144.58
MDL Number MFCD00000571
UN Number UN1993
Beilstein 1931682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1475314

 
alfa aesar™
A1475314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1475322

 
alfa aesar™
A1475322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1475330

 
alfa aesar™
A1475330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.58
Chemical Name or Material 4-Chloro-2-fluorotoluene
Assay 99%
Molecular Formula C7H6ClF
MDL Number MFCD00000571
CAS 452-75-5
UN Number UN1993
Beilstein 1931682
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 51°C (123°F)
Odor Aromatic
Boiling Point 158° to 160°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.