missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorotoluene, 99%

Manufacturer:  Alfa Aesar™ A1475330

 View more versions of this product

Catalog No. AAA1475330



Specifications

Specifications

4-Chloro-2-fluorotoluene
Aromatic
250g
C7H6ClF
UN1993
4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene, 4-chloro-2-fluorotoluene
CC1=C(C=C(C=C1)Cl)F
144.573
144.58
51°C (123°F)
158°C to 160°C
452-75-5
MFCD00000571
1931682
MKFCYQTVSDCXAQ-UHFFFAOYSA-N
4-chloro-2-fluoro-1-methylbenzene
520630
99%

Chemical Identifiers

452-75-5
144.573
MKFCYQTVSDCXAQ-UHFFFAOYSA-N
520630
CC1=C(C=C(C=C1)Cl)F
C7H6ClF
MFCD00000571
4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene, 4-chloro-2-fluorotoluene
4-chloro-2-fluoro-1-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents