missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-2-fluorophenylboronic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC447660050
Specifications
4-Chloro-2-fluorophenylboronic acid | |
289.2°C | |
5g | |
94% min. (HPLC) | |
2-fluoro-4-chlorophenylboronic acid, 4-chloro-2-fluoro-phenyl boronic acid, 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenylboronicacid, b-4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034 | |
B(C1=C(C=C(C=C1)Cl)F)(O)O | |
174.37 | |
174.37 |
128.7°C | |
233.5°C to 235.5°C | |
160591-91-3 | |
C6H5BClFO2 | |
YBNDRTRLXPEWKQ-UHFFFAOYSA-N | |
(4-chloro-2-fluorophenyl)boronic acid | |
4193595 | |
95% |
Chemical Identifiers
160591-91-3 | |
174.37 | |
2-fluoro-4-chlorophenylboronic acid, 4-chloro-2-fluoro-phenyl boronic acid, 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenylboronicacid, b-4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034 | |
(4-chloro-2-fluorophenyl)boronic acid |
C6H5BClFO2 | |
YBNDRTRLXPEWKQ-UHFFFAOYSA-N | |
4193595 | |
B(C1=C(C=C(C=C1)Cl)F)(O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning