Please login to your online account to display your discounted pricing

4-Chloro-2-fluorophenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-fluorophenylboronic acid
Assay Percent Range 95%
Molecular Formula C6H5BClFO2
Formula Weight 174.37
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC447660010

 
thermo fisher scientific
AC447660010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC447660050

 
thermo fisher scientific
AC447660050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 174.37
Quantity 1g
Molecular Formula C6H5BClFO2
Assay Percent Range 95%
CAS 160591-91-3
Chemical Name or Material 4-Chloro-2-fluorophenylboronic acid