missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-fluorophenylboronic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC447660010

 View more versions of this product

Catalog No. AC447660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chloro-2-fluorophenylboronic acid
Glass Bottle
233.5°C to 235.5°C
160591-91-3
C6H5BClFO2
YBNDRTRLXPEWKQ-UHFFFAOYSA-N
(4-chloro-2-fluorophenyl)boronic acid
4193595
95%
128.7°C
289.2°C
1g
94% min. (HPLC)
2-fluoro-4-chlorophenylboronic acid, 4-chloro-2-fluoro-phenyl boronic acid, 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenylboronicacid, b-4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034
B(C1=C(C=C(C=C1)Cl)F)(O)O
174.37
174.37

Chemical Identifiers

160591-91-3
174.37
2-fluoro-4-chlorophenylboronic acid, 4-chloro-2-fluoro-phenyl boronic acid, 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenylboronicacid, b-4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034
(4-chloro-2-fluorophenyl)boronic acid
C6H5BClFO2
YBNDRTRLXPEWKQ-UHFFFAOYSA-N
4193595
B(C1=C(C=C(C=C1)Cl)F)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents