Please login to your online account to display your discounted pricing

4-Chloro-2-fluorophenol, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-fluorophenol
CAS 348-62-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H4ClFO
Linear Formula ClC6H3FOH
Formula Weight 146.55
MDL Number MFCD00075030
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378210050

  SDS
fisher scientific acros
AC378210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC378210250

  SDS
thermo fisher scientific
AC378210250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid or Low Melting Solid
Formula Weight 146.55
Quantity 5g
Color Colorless to pink
Density 1.3780g/mL
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 348-62-9
Packaging Glass bottle
Linear Formula ClC6H3FOH
Molecular Formula C6H4ClFO
Assay Percent Range 98+%
Chemical Name or Material 4-Chloro-2-fluorophenol
MDL Number MFCD00075030
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.