missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorophenol, 98%

$112.82 - $371.26

Chemical Identifiers

CAS 348-62-9
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD00075030
InChI Key ZKMUKBBWORLNLA-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorophenol
PubChem CID 181975
IUPAC Name 4-chloro-2-fluorophenol
SMILES C1=CC(=C(C=C1Cl)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6141606 View Documents Alfa Aesar™
H6141606
5g Each for $112.82
 
AAH6141614 View Documents Alfa Aesar™
H6141614
25g Each for $371.26
 
Specifications

Chemical Identifiers

348-62-9
146.545
ZKMUKBBWORLNLA-UHFFFAOYSA-N
181975
C1=CC(=C(C=C1Cl)F)O
C6H4ClFO
MFCD00075030
2-fluoro-4-chlorophenol
4-chloro-2-fluorophenol

Specifications

1.378 g/mL
1.536
Colorless to Yellow
5g
C6H4ClFO
2-fluoro-4-chlorophenol
C1=CC(=C(C=C1Cl)F)O
146.545
146.55
98%
81°C (178°F)
103°C to 104°C
20°C
348-62-9
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
4-chloro-2-fluorophenol
181975
Liquid
4-Chloro-2-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures