Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorobenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-fluorobenzyl bromide
CAS 71916-82-0
Molecular Formula C7H5BrClF
Formula Weight 223.48
MDL Number MFCD00831105
UN Number UN3265
Synonym alpha-Bromo-4-chloro-2-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2681403

  SDS
alfa aesar™
H2681403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2681409

  SDS
alfa aesar™
H2681409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.48
Health Hazard 1 Danger
UN Number UN3265
Synonym alpha-Bromo-4-chloro-2-fluorotoluene
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 97%
Boiling Point 96° to 98°C (15mmHg)
Chemical Name or Material 4-Chloro-2-fluorobenzyl bromide
CAS 71916-82-0
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD00831105
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Molecular Formula C7H5BrClF
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.