missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6235103

 View more versions of this product

Catalog No. AAH6235103



Specifications

Specifications

4-Chloro-2-fluorobenzeneboronic acid pinacol ester
765917-27-9
1,3,2-dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chloro-2-fluorophenylboronic aicd, pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)Cl)F
256.508
256.51
96%
1g
C12H15BClFO2
HWHYSIMGKXYKDS-UHFFFAOYSA-N
2-(4-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22273588
Solid

Chemical Identifiers

765917-27-9
256.508
1,3,2-dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chloro-2-fluorophenylboronic aicd, pinacol ester
2-(4-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H15BClFO2
HWHYSIMGKXYKDS-UHFFFAOYSA-N
22273588
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)Cl)F
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents