missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorobenzeneboronic acid pinacol ester, 96%

$139.97 - $417.88

Chemical Identifiers

CAS 765917-27-9
Molecular Formula C12H15BClFO2
Molecular Weight (g/mol) 256.508
InChI Key HWHYSIMGKXYKDS-UHFFFAOYSA-N
Synonym 1,3,2-dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl
PubChem CID 22273588
IUPAC Name 2-(4-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62351MD View Documents Alfa Aesar™
H62351MD
250mg Each for $139.97
 
AAH6235103 View Documents Alfa Aesar™
H6235103
1g Each for $417.88
 
Specifications

Chemical Identifiers

765917-27-9
256.508
1,3,2-dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl
2-(4-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H15BClFO2
HWHYSIMGKXYKDS-UHFFFAOYSA-N
22273588
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)Cl)F

Specifications

250mg
C12H15BClFO2
HWHYSIMGKXYKDS-UHFFFAOYSA-N
2-(4-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22273588
Solid
4-Chloro-2-fluorobenzeneboronic acid pinacol ester
765917-27-9
1,3,2-dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)Cl)F
256.508
256.51
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures