missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-fluorobenzaldehyde, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC290620250

 View more versions of this product

Catalog No. AC290620250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chloro-2-fluorobenzaldehyde
Glass bottle
58.0°C to 61.0°C
61072-56-8
C7H4ClFO
2-fluoro-4-chlorobenzaldehyde, 3-fluoro-4-formylchlorobenzene, 4-chloranyl-2-fluoranyl-benzaldehyde, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, acmc-1b5ai, benzaldehyde, 4-chloro-2-fluoro, ksc494c2f, pubchem1435, timtec-bb sbb003985
UVGYSEIWAOOIJR-UHFFFAOYSA-N
4-chloro-2-fluorobenzaldehyde
2724908
Crystals or Powder
Authentic
White to Yellow
25g
98.5% min. (GC)
ClC6H3(F)CHO
Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
C1=CC(=C(C=C1Cl)F)C=O
158.56
158.56
99%

Chemical Identifiers

61072-56-8
158.56
2-fluoro-4-chlorobenzaldehyde, 3-fluoro-4-formylchlorobenzene, 4-chloranyl-2-fluoranyl-benzaldehyde, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, acmc-1b5ai, benzaldehyde, 4-chloro-2-fluoro, ksc494c2f, pubchem1435, timtec-bb sbb003985
4-chloro-2-fluorobenzaldehyde
C7H4ClFO
UVGYSEIWAOOIJR-UHFFFAOYSA-N
2724908
C1=CC(=C(C=C1Cl)F)C=O
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents