Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluorobenzaldehyde, 98%

$521.34 - $521.34

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-fluorobenzaldehyde
CAS 61072-56-8
Molecular Formula C7H4ClFO
Formula Weight 158.56
MDL Number MFCD00143282
Beilstein 3537705
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1611009

  SDS
alfa aesar™
A1611009
10g Each for $521.34
Description & Specifications

Specifications

Formula Weight 158.56
Quantity 10g
Chemical Name or Material 4-Chloro-2-fluorobenzaldehyde
Melting Point 58° to 60°C
Beilstein 3537705
Molecular Formula C7H4ClFO
CAS 61072-56-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00143282
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%

Applications
4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.