Please login to your online account to display your discounted pricing

4'-Chloro-2'-fluoroacetanilide, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  249450050

 View more versions of this product

Catalog No. AC249450050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 187.6
Physical Form Crystalline Powder
Quantity 5g
Color Pink to light brown
Melting Point 155° to 158°C
TSCA TSCA
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 4';-Chloro-2';-fluoroacetanilide
CAS 59280-70-5
Linear Formula NHC(O)CH3C6H3ClF
Assay Percent Range 99+%
MDL Number MFCD00042520
Name Note 99+%
Molecular Formula C8H7ClFNO
Packaging Glass bottle
Synonym N-(4-Chloro-2-fluorophenyl)acetamide
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4';-Chloro-2';-fluoroacetanilide
Name Note 99+%
CAS 59280-70-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H7ClFNO
Linear Formula NHC(O)CH3C6H3ClF
Formula Weight 187.6
MDL Number MFCD00042520
Synonym N-(4-Chloro-2-fluorophenyl)acetamide
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.

SDS & Safety Documents