Please login to your online account to display your discounted pricing

4'-Chloro-2'-fluoroacetanilide, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4';-Chloro-2';-fluoroacetanilide
Name Note 99+%
CAS 59280-70-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H7ClFNO
Linear Formula NHC(O)CH3C6H3ClF
Formula Weight 187.6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC249450050

  SDS
thermo fisher scientific
249450050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 187.6
Physical Form Crystalline Powder
Quantity 5g
Color Pink to light brown
Melting Point 155° to 158°C
TSCA TSCA
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 4';-Chloro-2';-fluoroacetanilide
CAS 59280-70-5
Linear Formula NHC(O)CH3C6H3ClF
Assay Percent Range 99+%
MDL Number MFCD00042520
Name Note 99+%
Molecular Formula C8H7ClFNO
Packaging Glass bottle
Synonym N-(4-Chloro-2-fluorophenyl)acetamide
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.