missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-2-fluoro-5-nitrotoluene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6184106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chloro-2-fluoro-5-nitrotoluene | |
42°C to 45°C | |
C7H5ClFNO2 | |
5 g | |
SJDPAVRCQNFVDM-UHFFFAOYSA-N | |
1-chloro-5-fluoro-4-methyl-2-nitrobenzene | |
87593 | |
98% |
18349-11-6 | |
113°C (235°F) | |
MFCD00134231 | |
4-chloro-2-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-4-methyl-2-nitro-benzene, toluene, 4-chloro-2-fluoro-5-nitro, benzene,1-chloro-5-fluoro-4-methyl-2-nitro, benzene, 1-chloro-5-fluoro-4-methyl-2-nitro, acmc-1c7av, 4-chloro-2-fluoro-5-nitro-toluene, buttpark 146\15-31, 2-chloro-4-floro-5-methylnitrobenzene, 2-chloro-4-fluoro-5-methylnitrobenzene | |
CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O | |
189.57 | |
189.57 |
Chemical Identifiers
18349-11-6 | |
189.57 | |
SJDPAVRCQNFVDM-UHFFFAOYSA-N | |
1-chloro-5-fluoro-4-methyl-2-nitrobenzene |
C7H5ClFNO2 | |
MFCD00134231 | |
87593 | |
CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 242-225-9
TSCA : No
Recommended Storage : Ambient temperatures