missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluoro-5-nitrobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ H6376006

 View more versions of this product

Catalog No. AAH6376006



Specifications

Specifications

4-Chloro-2-fluoro-5-nitrobenzoic acid
35112-05-1
MFCD04115725
KAONECPATVTFST-UHFFFAOYSA-N
219.552
219.55
5g
C7H3ClFNO4
4-chloro-2-fluoro-5-nitrobenzoic acid, 4-chloro-2-fluoro-5-nitrobenzoicacid, 4-chloro-2-fluoro-5-nitro-benzoic acid, 2-fluoro-4-chloro-5-nitrobenzoic acid, benzoic acid, 4-chloro-2-fluoro-5-nitro, 4-chloro-2-fluoro-5-nitrobenzoicacid, pubchem22286, acmc-1ah9o, ksc573m6p, 5-carboxy-2-chloro-4-fluoronitrobenzene
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)F)C(=O)O
3774553
98%

Chemical Identifiers

35112-05-1
219.552
KAONECPATVTFST-UHFFFAOYSA-N
3774553
C7H3ClFNO4
MFCD04115725
4-chloro-2-fluoro-5-nitrobenzoic acid, 4-chloro-2-fluoro-5-nitrobenzoicacid, 4-chloro-2-fluoro-5-nitro-benzoic acid, 2-fluoro-4-chloro-5-nitrobenzoic acid, benzoic acid, 4-chloro-2-fluoro-5-nitro, 4-chloro-2-fluoro-5-nitrobenzoicacid, pubchem22286, acmc-1ah9o, ksc573m6p, 5-carboxy-2-chloro-4-fluoronitrobenzene
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)F)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents