missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluoro-3-methoxybenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H3311406

 View more versions of this product

Catalog No. AAH3311406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chloro-2-fluoro-3-methoxybenzoic acid
1169870-80-7
MFCD19687204
COC1=C(C=CC(=C1F)C(=O)O)Cl
204.581
204.59
97%
5g
C8H6ClFO3
ODAJPJWSLSBLLM-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxybenzoic acid
57782418
Solid

Chemical Identifiers

1169870-80-7
204.581
ODAJPJWSLSBLLM-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxybenzoic acid
C8H6ClFO3
MFCD19687204
57782418
COC1=C(C=CC(=C1F)C(=O)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents