Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine
Formula Weight 247.65
MDL Number MFCD19687211
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3414503

 
alfa aesar™
H3414503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH34145MD

 
alfa aesar™
H34145MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.65
Chemical Name or Material 4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine
Physical Form Solid
Assay 97%
MDL Number MFCD19687211
Quantity 1g