Please login to your online account to display your discounted pricing

4-Chloro-2'-butyrothienone 97%, Acros Organics


Quantity 50mL

Manufacturer: thermo fisher scientific  154640500

 View more versions of this product

Catalog No. AC154640500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 188.67
Quantity 50mL
Color Clear gold to dark colored
EINECS Number 256-076-2
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Chloro-2'-butyrothienone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H9ClOS
MDL Number MFCD00005447
CAS 43076-59-1
Refractive Index 1.5635 to 1.5685 (20°C, 589nm)
Synonym 4-Chloro-1-(2-thienyl)-1-butanone
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Chloro-2'-butyrothienone
Name Note 97%
CAS 43076-59-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9ClOS
Formula Weight 188.67
MDL Number MFCD00005447
Synonym 4-Chloro-1-(2-thienyl)-1-butanone