missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2'-butyrothienone 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 154640100

 View more versions of this product

Catalog No. AC154640100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Chloro-2'-butyrothienone
10mL
43076-59-1
100.0
1.5635 to 1.5685 (20°C, 589nm)
≥96.0%
C8H9ClOS
MFCD00005447
Clear gold to dark colored
97%
96.0
Authentic
Liquid
97%
188.67
4-Chloro-1-(2-thienyl)-1-butanone

Chemical Identifiers

43076-59-1
100.0
C8H9ClOS
MFCD00005447
4-Chloro-2'-butyrothienone, 97%
96.0
97%
188.67
4-Chloro-1-(2-thienyl)-1-butanone
≥96.0%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 256-076-2