Please login to your online account to display your discounted pricing

4-Chloro-2'-butyrothienone 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Chloro-2'-butyrothienone
Name Note 97%
CAS 43076-59-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9ClOS
Formula Weight 188.67
MDL Number MFCD00005447
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC154640100

  SDS
thermo fisher scientific
154640100
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC154640500

  SDS
thermo fisher scientific
154640500
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 188.67
Quantity 10mL
Color Clear gold to dark colored
EINECS Number 256-076-2
CAS Min % 96.0
CAS Max % 100.0
CAS 43076-59-1
MDL Number MFCD00005447
Chemical Name or Material 4-Chloro-2'-butyrothienone
Assay Percent Range 97%
Molecular Formula C8H9ClOS
Name Note 97%
Refractive Index 1.5635 to 1.5685 (20°C, 589nm)
Synonym 4-Chloro-1-(2-thienyl)-1-butanone
Infrared Spectrum Authentic