Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,8-dimethylquinoline

$110.82 - $384.82

Chemical Identifiers

Chemical Name or Material 4-Chloro-2,8-dimethylquinoline
CAS 32314-39-9
Molecular Formula C11H10ClN
Formula Weight 191.66
MDL Number MFCD00272394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5171403

 
alfa aesar™
H5171403
1g Each for $384.82

AAH51714MD

 
alfa aesar™
H51714MD
250mg Each for $110.82
Description & Specifications

Specifications

Formula Weight 191.66
Chemical Name or Material 4-Chloro-2,8-dimethylquinoline
Molecular Formula C11H10ClN
MDL Number MFCD00272394
CAS 32314-39-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.