Please login to your online account to display your discounted pricing

4-Chloro-2,8-bis(trifluoromethyl)quinoline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2, 8-bis(trifluoromethyl)quinoline
Name Note 97%
CAS 83012-13-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H4ClF6N
Formula Weight 299.6
MDL Number MFCD00075104
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC306020010

  SDS
thermo fisher scientific
AC306020010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC306022500

  SDS
thermo fisher scientific
306022500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 299.6
Quantity 1g
Color Beige
Melting Point 45° to 49°C
EINECS Number 280-132-5
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 4-Chloro-2, 8-bis(trifluoromethyl)quinoline
Assay Percent Range 97%
Molecular Formula C11H4ClF6N
MDL Number MFCD00075104
CAS 83012-13-9
Name Note 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.