Please login to your online account to display your discounted pricing

4-Chloro-2,8-bis(trifluoromethyl)quinoline, 97%, ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  306022500

 View more versions of this product

Catalog No. AC306022500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 299.6
Quantity 250mg
Color Beige
EINECS Number 280-132-5
Melting Point 45° to 49°C
CAS Min % 96.0
CAS Max % 100.0
Name Note 97%
Molecular Formula C11H4ClF6N
Chemical Name or Material 4-Chloro-2, 8-bis(trifluoromethyl)quinoline
Assay Percent Range 97%
CAS 83012-13-9
MDL Number MFCD00075104
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chloro-2, 8-bis(trifluoromethyl)quinoline
Name Note 97%
CAS 83012-13-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H4ClF6N
Formula Weight 299.6
MDL Number MFCD00075104
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents