missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,6-dimethylpyrimidine, 97%

Manufacturer:  Alfa Aesar™ H5473506

 View more versions of this product

Catalog No. AAH5473506



Specifications

Specifications

4-Chloro-2,6-dimethylpyrimidine
4472-45-1
MFCD00234197
GSXFOGXQLRLSKK-UHFFFAOYSA-N
4-chloro-2,6-dimethylpyrimidine
3154199
97%
5g
C6H7ClN2
2,4-dimethyl-6-chloropyrimidine
CC1=CC(=NC(=N1)C)Cl
142.586
142.03

Chemical Identifiers

4472-45-1
142.586
GSXFOGXQLRLSKK-UHFFFAOYSA-N
3154199
CC1=CC(=NC(=N1)C)Cl
C6H7ClN2
MFCD00234197
2,4-dimethyl-6-chloropyrimidine
4-chloro-2,6-dimethylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents