Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,6-dimethyl-3-nitropyridine, 98%

Chemical Identifiers

Chemical Name or Material 4-Chloro-2,6-dimethyl-3-nitropyridine
CAS 15513-48-1
Formula Weight 186.6
MDL Number MFCD08543468
Beilstein 1569403
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6418006

  SDS
alfa aesar™
H6418006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6418003

  SDS
alfa aesar™
H6418003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64180MD

  SDS
alfa aesar™
H64180MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 70° to 73°C
Formula Weight 186.6
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Off-white
Chemical Name or Material 4-Chloro-2,6-dimethyl-3-nitropyridine
Beilstein 1569403
Storage Note 1 2° to 8°C
Quantity 5g
Health Hazard 1 Danger
CAS 15513-48-1
Physical Form Solid
MDL Number MFCD08543468
Assay 98%
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C