missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,6-dimethyl-3-nitropyridine, 98%

Manufacturer:  Alfa Aesar™ H6418006

 View more versions of this product

Catalog No. AAH6418006



Specifications

Specifications

4-Chloro-2,6-dimethyl-3-nitropyridine
70°C to 73°C
15513-48-1
MFCD08543468
pyridine, 4-chloro-2,6-dimethyl-3-nitro, 4-chloro-3-nitro-2,6-lutidine, pubchem11177, 4-chloranyl-2,6-dimethyl-3-nitro-pyridine
CC1=NC(=C(C(=C1)Cl)[N+](=O)[O-])C
186.595
186.6
98%
White
5g
C7H7ClN2O2
1569403
PSBGFLWVQDGETK-UHFFFAOYSA-N
4-chloro-2,6-dimethyl-3-nitropyridine
10702725
Solid

Chemical Identifiers

15513-48-1
186.595
PSBGFLWVQDGETK-UHFFFAOYSA-N
10702725
CC1=NC(=C(C(=C1)Cl)[N+](=O)[O-])C
C7H7ClN2O2
MFCD08543468
pyridine, 4-chloro-2,6-dimethyl-3-nitro, 4-chloro-3-nitro-2,6-lutidine, pubchem11177, 4-chloranyl-2,6-dimethyl-3-nitro-pyridine
4-chloro-2,6-dimethyl-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : 2° to 8°C

SDS Documents