Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,6-difluorobenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-2,6-difluorobenzyl bromide
CAS 537013-52-8
Molecular Formula C7H4BrClF2
Formula Weight 241.46
MDL Number MFCD03094463
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670703

 
alfa aesar™
H2670703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2670706

 
alfa aesar™
H2670706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 241.46
Chemical Name or Material 4-Chloro-2,6-difluorobenzyl bromide
Assay 97%
Molecular Formula C7H4BrClF2
MDL Number MFCD03094463
CAS 537013-52-8
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Used frequently in organic synthesis. Find application as propellants.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.