Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,6-difluorobenzaldehyde, 97%

$147.35 - $1,118.01

Chemical Identifiers

Chemical Name or Material 4-Chloro-2,6-difluorobenzaldehyde
CAS 252004-45-8
Molecular Formula C7H3ClF2O
Formula Weight 176.55
MDL Number MFCD03094460
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2645503

  SDS
alfa aesar™
H2645503
1g Each for $147.35

AAH2645509

  SDS
alfa aesar™
H2645509
10g Each for $1,118.01
Description & Specifications

Specifications

Formula Weight 176.55
Chemical Name or Material 4-Chloro-2,6-difluorobenzaldehyde
Assay 97%
Molecular Formula C7H3ClF2O
MDL Number MFCD03094460
CAS 252004-45-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 70° to 74°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.