Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chloro-2',6'-difluoroacetophenone, 97%

$121.91 - $479.15

Chemical Identifiers

Chemical Name or Material 4'-Chloro-2',6'-difluoroacetophenone
CAS 1017777-45-5
Molecular Formula C8H5ClF2O
Formula Weight 190.58
MDL Number MFCD09832259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3271103

  SDS
alfa aesar™
H3271103
1g Each for $121.91

AAH3271106

  SDS
alfa aesar™
H3271106
5g Each for $479.15
Description & Specifications

Specifications

Formula Weight 190.58
Molecular Formula C8H5ClF2O
MDL Number MFCD09832259
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 4'-Chloro-2',6'-difluoroacetophenone
Assay 97%
CAS 1017777-45-5
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.