Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-(5-isoxazolyl)phenol, 97%

$28.49 - $307.38

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-(5-isoxazolyl)phenol
CAS 86176-56-9
Molecular Formula C9H6ClNO2
Formula Weight 195.61
MDL Number MFCD00218660
Synonym 5-(5-Chloro-2-hydroxyphenyl)isoxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2733303

  SDS
alfa aesar™
H2733303
1g Each for $28.49

AAH2733306

  SDS
alfa aesar™
H2733306
5g Each for $86.78

AAH2733314

  SDS
alfa aesar™
H2733314
25g Each for $307.38
Description & Specifications

Specifications

Formula Weight 195.61
Chemical Name or Material 4-Chloro-2-(5-isoxazolyl)phenol
Assay 97%
Molecular Formula C9H6ClNO2
MDL Number MFCD00218660
CAS 86176-56-9
Synonym 5-(5-Chloro-2-hydroxyphenyl)isoxazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 200° to 204°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.