Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,5-dimethylbenzenesulfonamide, 97%

$159.72 - $436.96

Chemical Identifiers

Chemical Name or Material 4-Chloro-2,5-dimethylbenzenesulfonamide
CAS 219689-73-3
Molecular Formula C8H10ClNO2S
Formula Weight 219.69
MDL Number MFCD00275263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2168303

  SDS
alfa aesar™
B2168303
1g Each for $159.72

AAB2168306

  SDS
alfa aesar™
B2168306
5g Each for $436.96
Description & Specifications

Specifications

Formula Weight 219.69
MDL Number MFCD00275263
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Health Hazard 1 Warning
CAS 219689-73-3
Chemical Name or Material 4-Chloro-2,5-dimethylbenzenesulfonamide
Quantity 1g
Melting Point 190° to 192°C
Molecular Formula C8H10ClNO2S
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.