missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,5-difluorobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ H5552106

 View more versions of this product

Catalog No. AAH5552106



Specifications

Specifications

4-Chloro-2,5-difluorobenzoic acid
5g
C7H3ClF2O2
2,5-difluoro-4-chloro-benzoic acid
C1=C(C(=CC(=C1F)Cl)F)C(=O)O
192.546
192.55
154°C to 157°C
132794-07-1
MFCD00077480
PEPCYJSDHYMIFN-UHFFFAOYSA-N
4-chloro-2,5-difluorobenzoic acid
2733274
98%

Chemical Identifiers

132794-07-1
192.546
PEPCYJSDHYMIFN-UHFFFAOYSA-N
2733274
C1=C(C(=CC(=C1F)Cl)F)C(=O)O
C7H3ClF2O2
MFCD00077480
2,5-difluoro-4-chloro-benzoic acid
4-chloro-2,5-difluorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

TSCA : No

Recommended Storage : Ambient temperatures

Documents