missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,5-difluorobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ H5552114

 View more versions of this product

Catalog No. AAH5552114



Specifications

Specifications

4-Chloro-2,5-difluorobenzoic acid
25g
C7H3ClF2O2
2,5-difluoro-4-chloro-benzoic acid
C1=C(C(=CC(=C1F)Cl)F)C(=O)O
192.546
192.55
154°C to 157°C
132794-07-1
MFCD00077480
PEPCYJSDHYMIFN-UHFFFAOYSA-N
4-chloro-2,5-difluorobenzoic acid
2733274
98%

Chemical Identifiers

132794-07-1
192.546
PEPCYJSDHYMIFN-UHFFFAOYSA-N
2733274
C1=C(C(=CC(=C1F)Cl)F)C(=O)O
C7H3ClF2O2
MFCD00077480
2,5-difluoro-4-chloro-benzoic acid
4-chloro-2,5-difluorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents