Please login to your online account to display your discounted pricing

4-Chloro-2,5-difluorobenzoic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC309690010

 View more versions of this product

Catalog No. AC309690010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 192.55
Quantity 1g
Color Off-white
Melting Point 153° to 158°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00077480
Assay Percent Range 97%
Name Note 97%
Molecular Formula C7H3ClF2O2
Linear Formula ClC6HF(COOH)
Chemical Name or Material 4-Chloro-2, 5-difluorobenzoic acid
CAS 132794-07-1
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.

Specifications

Chemical Name or Material 4-Chloro-2, 5-difluorobenzoic acid
Name Note 97%
CAS 132794-07-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H3ClF2O2
Linear Formula ClC6HF(COOH)
Formula Weight 192.55
MDL Number MFCD00077480
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.

SDS & Safety Documents