missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,5-difluorobenzaldehyde, 98%

$108.41 - $518.15

Chemical Identifiers

CAS 879093-02-4
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD07782051
InChI Key XPZYVMBPIANKJG-UHFFFAOYSA-N
Synonym 4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
PubChem CID 18991142
IUPAC Name 4-chloro-2,5-difluorobenzaldehyde
SMILES C1=C(C(=CC(=C1F)Cl)F)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64112MD View Documents Alfa Aesar™
H64112MD
250mg Each for $108.41
 
AAH6411203 View Documents Alfa Aesar™
H6411203
1g Each for $259.08
 
AAH6411206 View Documents Alfa Aesar™
H6411206
5g Each for $518.15
 
Specifications

Chemical Identifiers

879093-02-4
176.547
XPZYVMBPIANKJG-UHFFFAOYSA-N
18991142
C1=C(C(=CC(=C1F)Cl)F)C=O
C7H3ClF2O
MFCD07782051
4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
4-chloro-2,5-difluorobenzaldehyde

Specifications

White
879093-02-4
MFCD07782051
4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
C1=C(C(=CC(=C1F)Cl)F)C=O
176.547
176.55
98%
250mg
C7H3ClF2O
14516595
XPZYVMBPIANKJG-UHFFFAOYSA-N
4-chloro-2,5-difluorobenzaldehyde
18991142
Solid
4-Chloro-2,5-difluorobenzaldehyde