missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2,5-difluorobenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ H6411206

 View more versions of this product

Catalog No. AAH6411206



Specifications

Specifications

4-Chloro-2,5-difluorobenzaldehyde
5g
C7H3ClF2O
14516595
XPZYVMBPIANKJG-UHFFFAOYSA-N
4-chloro-2,5-difluorobenzaldehyde
18991142
Solid
White
879093-02-4
MFCD07782051
4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
C1=C(C(=CC(=C1F)Cl)F)C=O
176.547
176.55
98%

Chemical Identifiers

879093-02-4
176.547
XPZYVMBPIANKJG-UHFFFAOYSA-N
18991142
C1=C(C(=CC(=C1F)Cl)F)C=O
C7H3ClF2O
MFCD07782051
4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
4-chloro-2,5-difluorobenzaldehyde
Documents