missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-2,3-dimethylpyridine N-Oxide 98.0+%, TCI America™
$79.63 - $275.40
Chemical Identifiers
CAS | 59886-90-7 |
---|---|
Molecular Formula | C7H9ClNO |
Molecular Weight (g/mol) | 158.61 |
MDL Number | MFCD06411125 |
InChI Key | HHYDNQMGLBHVSW-UHFFFAOYNA-N |
PubChem CID | 9877446 |
IUPAC Name | 4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate |
SMILES | CC1=C(C)C(Cl)=C[CH-][NH++]1[O-] |
Chemical Identifiers
59886-90-7 | |
158.61 | |
HHYDNQMGLBHVSW-UHFFFAOYNA-N | |
4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate |
C7H9ClNO | |
MFCD06411125 | |
9877446 | |
CC1=C(C)C(Cl)=C[CH-][NH++]1[O-] |
Specifications
59886-90-7 | |
Yellow | |
MFCD06411125 | |
HHYDNQMGLBHVSW-UHFFFAOYNA-N | |
4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate | |
9877446 | |
≥98.0% (GC,T) | |
4-Chloro-2,3-dimethylpyridine N-Oxide |
108°C | |
C7H9ClNO | |
5 g | |
CC1=C(C)C(Cl)=C[CH-][NH++]1[O-] | |
158.61 | |
157.60 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer