Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-1H-pyrazolo[3,4-b]pyridine, 98%

Chemical Identifiers

Chemical Name or Material 4-Chloro-1H-pyrazolo[3,4-b]pyridine
CAS 29274-28-0
Formula Weight 153.57
MDL Number MFCD11846313
Beilstein 607874
Synonym 5-Chloro-2,8,9-triazabicyclo[4.3.0]nona-1,3,5,7-tetraene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66767MD

 
alfa aesar™
H66767MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.57
CAS 29274-28-0
Assay 98%
MDL Number MFCD11846313
Beilstein 607874
Synonym 5-Chloro-2,8,9-triazabicyclo[4.3.0]nona-1,3,5,7-tetraene
Chemical Name or Material 4-Chloro-1H-pyrazolo[3,4-b]pyridine
Physical Form Solid
Quantity 250mg