missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-1-naphthol 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 109551000

 View more versions of this product

Catalog No. AC109551000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

178.62
Powder or Needles
Off-white to gray to light brown
117° to 122°C
96.0
C10H7ClO
4-Chloro-1-naphthol
97%
Authentic
≥96.0%
100g
06, 611
100.0
ClC10H6OH
99%
604-44-4
MFCD00003974
Glass bottle

Chemical Identifiers

604-44-4
100.0
C10H7ClO
178.62
4-Chloro-1-naphthol, 99%
96.0
97%
ClC10H6OH
MFCD00003974
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-068-5

TSCA : TSCA