missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-1-methylpiperidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C1085100G
Specifications
4-Chloro-1-methylpiperidine Hydrochloride | |
162°C | |
C6H13Cl2N | |
100 g | |
QDOJNWFOCOWAPO-UHFFFAOYSA-N | |
4-chloro-1-methylpiperidine;hydrochloride | |
3034158 | |
≥98.0% (T) |
5382-23-0 | |
White-Yellow | |
MFCD00012834 | |
4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n | |
CN1CCC(CC1)Cl.Cl | |
170.077 | |
170.08 | |
Crystalline Powder |
Chemical Identifiers
5382-23-0 | |
170.077 | |
QDOJNWFOCOWAPO-UHFFFAOYSA-N | |
3034158 | |
CN1CCC(CC1)Cl.Cl |
C6H13Cl2N | |
MFCD00012834 | |
4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n | |
4-chloro-1-methylpiperidine;hydrochloride |
Safety and Handling
TSCA : No