Please login to your online account to display your discounted pricing

4-Chloro-1-methylpiperidine hydrochloride, 97%, Acros Organics


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  109495000

 View more versions of this product

Catalog No. AC109495000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 170.08
Physical Form Crystalline Powder
Quantity 500g
Color White to light beige
EINECS Number 226-375-2
Melting Point 160° to 165°C
CAS Max % 100.0
CAS Min % 96.0
Synonym 4-Chloro-N-methylpiperidine hydrochloride
Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride
Molecular Formula C6H12ClN·HCl
Assay Percent Range 97%
MDL Number MFCD00012834
Name Note 97%
CAS 5382-23-0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride
Name Note 97%
CAS 5382-23-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12ClN·HCl
Formula Weight 170.08
MDL Number MFCD00012834
Synonym 4-Chloro-N-methylpiperidine hydrochloride
Accessories

HYGROSCOPIC

SDS & Safety Documents