Please login to your online account to display your discounted pricing

4-Chloro-1-methylpiperidine hydrochloride, 97%, Acros Organics

$451.71 - $451.71

Chemical Identifiers

Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride
Name Note 97%
CAS 5382-23-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12ClN·HCl
Formula Weight 170.08
MDL Number MFCD00012834
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC109495000

  SDS
thermo fisher scientific
109495000
Glass bottle 500g Each for $451.71
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 170.08
Physical Form Crystalline Powder
Quantity 500g
Color White to light beige
EINECS Number 226-375-2
Melting Point 160° to 165°C
CAS Max % 100.0
CAS Min % 96.0
Synonym 4-Chloro-N-methylpiperidine hydrochloride
Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride
Molecular Formula C6H12ClN·HCl
Assay Percent Range 97%
MDL Number MFCD00012834
Name Note 97%
CAS 5382-23-0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Safety and Handling

HYGROSCOPIC