Please login to your online account to display your discounted pricing

4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde, 95+%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C5H5ClN2O
Formula Weight 144.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03946DA

 
fisher scientific company
RF03946DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.56
Quantity 1g
Assay 95%
Chemical Name or Material 4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Assay Percent Range 95%
Molecular Formula C5H5ClN2O
CAS 175204-81-6
Packaging Amber Glass Bottle