Please login to your online account to display your discounted pricing

4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde, 95+%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RF03946DA

 View more versions of this product

Catalog No. RF03946DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 144.56
Quantity 1g
Assay 95%
Chemical Name or Material 4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
CAS 175204-81-6
Packaging Amber Glass Bottle
Molecular Formula C5H5ClN2O
Assay Percent Range 95%

Specifications

Chemical Name or Material 4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C5H5ClN2O
Formula Weight 144.56