Please login to your online account to display your discounted pricing

4-Chloro-1,8-naphthalic anhydride, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-1, 8-naphthalic anhydride
Name Note tech.
CAS 4053-08-1
Purity Grade Technical Grade
Molecular Formula C12H5ClO3
Formula Weight 232.62
MDL Number MFCD00006928
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC184652500

  SDS
thermo fisher scientific
184652500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Powder
Formula Weight 232.62
Quantity 250g
Color Beige to brown
Beilstein 17, 522
EINECS Number 223-760-7
RTECS Number QL6127295
TSCA TSCA
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Chloro-1, 8-naphthalic anhydride
Name Note tech.
CAS 4053-08-1
Molecular Formula C12H5ClO3
MDL Number MFCD00006928
Infrared Spectrum Authentic
Loss on Drying 50% max.
Additional Information inorganic salts: < 2%
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.