missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444670050
Specifications
4-(Carboxymethyl)phenylboronic acid pinacol ester | |
White to Yellow | |
5g | |
97% | |
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylacetate, 4-carboxymethyl phenylboronic acid pinacol ester, 4-carboxymethyl phenylboronic acidpinacol ester, 4-dihydroxyborane phenylacetic acid pinacol ester, benzeneacetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, phenylacetic acid-4-boronic acid pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)O | |
262.11 | |
262.11 | |
97% |
96 % min. Consistent with structure (1H-NMR) | |
166°C to 168°C | |
797755-07-8 | |
C14H19BO4 | |
FNLWHBHWDXCWHV-UHFFFAOYSA-N | |
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid | |
3719002 | |
Crystalline Powder or Powder |
Chemical Identifiers
797755-07-8 | |
262.11 | |
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylacetate, 4-carboxymethyl phenylboronic acid pinacol ester, 4-carboxymethyl phenylboronic acidpinacol ester, 4-dihydroxyborane phenylacetic acid pinacol ester, benzeneacetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, phenylacetic acid-4-boronic acid pinacol ester | |
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid |
C14H19BO4 | |
FNLWHBHWDXCWHV-UHFFFAOYSA-N | |
3719002 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning